Anti-MSN Rabbit Polyclonal Antibody

Supplier: Proteintech
16495-1-AP-150UL
10090-516EA 402 USD
10090-516
Anti-MSN Rabbit Polyclonal Antibody
Antibodies
Western Blot:SGC-7901 Cells, 1:1000-1:10000; IHC: Human Placenta Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; FC: HepG2 Cells, N/A

Type: Primary
Antigen: MSN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR