Anti-IGHD Rabbit Polyclonal Antibody

Supplier: Proteintech
16489-1-AP-150UL
10088-668EA 402 USD
10088-668
Anti-IGHD Rabbit Polyclonal Antibody
Antibodies
IgD is the major antigen receptor isotype on the surface of most peripheral B-cells, where it is coexpressed with IgM. The membrane-bound IgD (mIgD) induces the phosphorylation of CD79A and CD79B by the Src family of protein tyrosine kinases. This antibody recognize the heavy chain (~60 kDa) of IgD.

Western Blot: Raji Cells, 1:200-1:2000; IHC: Human tonsillitis Tissue, 1:20-1:200

Type: Primary
Antigen: IGHD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR