Anti-PHAX Rabbit Polyclonal Antibody

Supplier: Proteintech
16481-1-AP-150UL
10092-178EA 402 USD
10092-178
Anti-PHAX Rabbit Polyclonal Antibody
Antibodies
Small nuclear and small nucleolar RNAs (snRNAs and snoRNAs) are essential components of snRNPs and snoRNPs, and have a critical role in the maturation of, respectively, mRNAs and rRNAs within the nucleus of eukaryotic cells. Complex and specific pathways exist for the assembly of snRNPs and snoRNPs, involving, nucleocytoplasmic transport of snRNAs and intranuclear transport between compartments of snoRNAs. In metazoa, a subset of spliceosomal snRNAs are exported from the nucleus after transcription. This export occurs in a large complex containing a snRNA, the nuclear cap binding complex (CBC), RanGTP, the leucine-rich nuclear export signal receptor CRM1/Xpo1, and the recently identified phosphoprotein PHAX (phosphorylated adaptor for RNA export). PHAX contains a conserved single-stranded nucleic acid binding domain (RNA_GG_bind domain) with no sequence homology with any other known RNA-binding module

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: PHAX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR