Anti-MVP Rabbit Polyclonal Antibody

Supplier: Proteintech
16478-1-AP-150UL
10090-668EA 402 USD
10090-668
Anti-MVP Rabbit Polyclonal Antibody
Antibodies
Vaults are the largest cellular ribonuclear protein complexes with a hollow barrel-like structure, and have been associated with the MDR phenotype. Major vault protein (MVP) is the main component of vaults and is presumed to be involved in MDR. MVP is identical with the human lung resistance protein (LRP), known to be overexpressed in multiple chemotherapy resistance models . MVP has been associated to resistance to radiotherapy [4], probably due to its role in preventing apoptosis by inhibiting the COP-1/p53 axis .

Western Blot:SH-SY5Y Cells, 1:500-1:5000; IHC: Human lung Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: MVP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR