Anti-ALB Rabbit Polyclonal Antibody

Supplier: Proteintech
16475-1-AP-150UL
10082-750EA 485.42 USD
10082-750
Anti-ALB Rabbit Polyclonal Antibody
Antibodies
Albumin is the most abundant protein in blood plasma. Alterations of level of serum albumin are linked to variety of diseases. Albumin is expressed exclusively by well-differentiated hepatocytes, thus anti-albumin has been used to mark hepatocytes. (21388516, 23832071)

Western Blot: Human Blood Tissue, 1:5000-1:50000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: ALB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR