Anti-PC6 Rabbit Polyclonal Antibody

Supplier: Proteintech
16470-1-AP-150UL
10091-900EA 402 USD
10091-900
Anti-PC6 Rabbit Polyclonal Antibody
Antibodies
PC5 ,a mammalian kex2-like processing endoprotease, mediates posttranslational endoproteolytic processing for several integrin alpha subunits.It has been identified in rat and mouse endocrine and neuroendocrine tissues,and is encoded by multiple mRNAs that generate two splice variant isoforms referred to as PC5A (102KDa) and PC5B (207KDa) .PC5A is smaller than PC5B and it can be processed into two forms (117 kDa and 65 kDa) in neuronal cells.

IHC: Human Brain Tissue, 1:20-1:200; Western Blot: Mouse Brain Tissue, 1:200-1:1000

Type: Primary
Antigen: PC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR