Anti-DPEP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
16466-1-AP-150UL
10086-088EA 402 USD
10086-088
Anti-DPEP2 Rabbit Polyclonal Antibody
Antibodies
DPEP2(Dipeptidase 2) may play an important role in the regulation of leukotriene activity which converts leukotriene d4 to leukotriene e4. It belongs to the peptidase M19 family. This protein has 2 isoforms produced by alternative splicing with the molecular weight of 53 kDa and 44 kDa.

Western Blot:Transfected HEK-293 Cells, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: DPEP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR