Anti-HGD Rabbit Polyclonal Antibody

Supplier: Proteintech
16465-1-AP-150UL
10088-174EA 402 USD
10088-174
Anti-HGD Rabbit Polyclonal Antibody
Antibodies
Homogentisate1,2-dioxygenase (HGD), also named as HGO, is a mononuclear Fe(II)-dependent oxygenase that catalyzes the third step in the pathway for the catabolism of tyrosine, the conversion of homogentisate (HG) to maleylacetoacetate (MAA) and it can exsit as a dimer or trimer. HGD consists of a single type of subunit with no intermolecular disulfide bridges and requires Fe2+ as a cofactor. Defects in HGD are the cause of alkaptonuria (AKU).

Western Blot: Human Liver Tissue, 1:500-1:5000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: HGD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR