Anti-CLDN3 Rabbit Polyclonal Antibody

Supplier: Proteintech
16456-1-AP-150UL
10084-832EA 402 USD
10084-832
Anti-CLDN3 Rabbit Polyclonal Antibody
Antibodies
Claudins are a family of proteins that are the most important components of the tight junctions, where they establish the paracellular barrier that controls the flow of molecules in the intercellular space between the cells of an epithelium. 23 claudins have been identified. They are small (20-27 kilodalton (kDa)) proteins with very similar structure. They have four transmembrane domains, with the N-terminus and the C-terminus in the cytoplasm.

Western Blot: Mouse large intestine Tissue, 1:200-1:1000

Type: Primary
Antigen: CLDN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR