Anti-AP3D1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16454-1-AP-150UL
10082-970EA 402 USD
10082-970
Anti-AP3D1 Rabbit Polyclonal Antibody
Antibodies
Adaptor protein (AP) complexes are cytosolic heterotetramers that mediate the sorting of membrane proteins in the secretory and endocytic pathways. AP3D1 is a subunit of the AP-3 complex which is composed of two large adaptins (AP3D1 and AP3B1 or AP3B2), a medium adaptin (AP3M1 or AP3M2) and a small adaptin (APS1 or AP3S2). AP-3 complex is associated with the Golgi region as well as more peripheral structures. It facilitates the budding of vesicles from the Golgi membrane and may be directly involved in trafficking to lysosomes.

Western Blot:NIH/3T3 Cells, 1:200-1:2000

Type: Primary
Antigen: AP3D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR