Anti-DEK Rabbit Polyclonal Antibody

Supplier: Proteintech
16448-1-AP-150UL
10085-390EA 402 USD
10085-390
Anti-DEK Rabbit Polyclonal Antibody
Antibodies
Dek oncogene(DEK) is first recognized in a fusion with CAN nucleoporin protein, and it's a also autoantigen in patients with pquciarticular onset juvenile rheumatoid arthritis. As a site-specific binding protein, DEK may involve in transcriptinoal regulaton and signal transdcution. Analysis by agarose gel celctrophoresis, DEK showed to reduce the efficiency with which chromation is replicated. It's one of the first components of the splicing complex, which remains associate with spliced exons dependent on prior splicing of pre-mRNA.

Western Blot: HeLa Cells, 1:1000-1:10000; IHC: Human Brain Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: DEK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR