Anti-CAV1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16447-1-AP-150UL
10084-116EA 537.7 USD
10084-116
Anti-CAV1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A549 Cells, 1:500-1:5000; IHC: Human lung Tissue, 1:20-1:200; IP:A549 Cells, 1:200-1:2000

Type: Primary
Antigen: CAV1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR