Anti-Carboxypeptidase M Rabbit Polyclonal Antibody

Supplier: Proteintech
16440-1-AP-150UL
10085-118EA 402 USD
10085-118
Anti-Carboxypeptidase M Rabbit Polyclonal Antibody
Antibodies
CPM(Carboxypeptidase M), a plasma membrane-bound enzyme, is present in many human organs and differs from other carboxypeptidases that cleave basic C-terminal amino acids. CPM is structurally, catalytically and immunologically distinct from other carboxypeptidases, and expressed on cell membranes in a glycosyl-phosphatidylinositol-anchored form and it can be a novel marker and cellular player in lipid uptake and/or metabolism of MAs by promoting foam cell formation . This full length protein has a signal peptide with 17 amino acid, a propeptide with 20 amino acid and five glycosylation sites.

Western Blot: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: Carboxypeptidase M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR