Anti-NELFB Rabbit Polyclonal Antibody

Supplier: Proteintech
16418-1-AP-150UL
10091-026EA 402 USD
10091-026
Anti-NELFB Rabbit Polyclonal Antibody
Antibodies
Western Blot:BxPC-3 Cells, 1:500-1:5000; IP: Mouse Brain Tissue, 1:200-1:2000; IF:BxPC-3 Cells, 1:10-1:100

Type: Primary
Antigen: NELFB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR