Anti-Antithrombin III Rabbit Polyclonal Antibody

Supplier: Proteintech
16414-1-AP-150UL
10093-566EA 402 USD
10093-566
Anti-Antithrombin III Rabbit Polyclonal Antibody
Antibodies
SERPINC1, also named Antithrombin III or Antithrombin, is a plasma protease inhibitor and a member of the serpin superfamily. This protein inhibits thrombin as well as other activated serine proteases of the coagulation system, and it regulates the blood coagulation cascade. The protein includes two functional domains: the heparin binding-domain at the N-terminus of the mature protein, and the reactive site domain at the C-terminus. The inhibitory activity is enhanced by the presence of heparin. More than 120 mutations have been identified for this gene, many of which are known to cause antithrombin-III deficiency.

Western Blot: Human Blood Tissue, 1:200-1:1000

Type: Primary
Antigen: Antithrombin III
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR