Anti-RPL23A Rabbit Polyclonal Antibody

Supplier: Proteintech
16386-1-AP-150UL
10093-536EA 524.48 USD
10093-536
Anti-RPL23A Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:200-1:2000; IHC: Human Liver, 1:20-1:200

Type: Primary
Antigen: RPL23A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR