Anti-WAPL; WAPAL Rabbit Polyclonal Antibody

Supplier: Proteintech
16370-1-AP-150UL
10097-060EA 402 USD
10097-060
Anti-WAPL; WAPAL Rabbit Polyclonal Antibody
Antibodies
The wings apart-like (WAPL, or WAPAL) protein was originally identified as a gene product that potentially regulates heterochromatin organization in Drosophila melanogaster. Human WAPL is a cohesin-binding protein that promotes sister-chromatid resolution in mitotic prophase. Overexpression of WAPL disturbs mitosis and cytokinesis, and contributes to tumor progression by induction of chromosomal instability.

Western Blot: HL-60 Cells, 1:500-1:5000

Type: Primary
Antigen: WAPL; WAPAL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR