Anti-FZR1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16368-1-AP-150UL
10087-208EA 537.7 USD
10087-208
Anti-FZR1 Rabbit Polyclonal Antibody
Antibodies
FZR1 gene, also named as FYR, FZR and KIAA1242, has been previously shown to be mutated in lobular breast carcinomas. Direct evidence of FZR1 mutations triggering tumorigenesis has been investigated. FZR1 is a key regulator of ligase activity of the anaphase promoting complex/cyclosome (APC/C) which is playing vital role s in cell cycle progression. There are 7 WD-repeat domains in FZR1. Upon phosphorylation and dephosphorylation, it interacts with other components to modulate cell mitosis. This gene is different to E-cadherin (CDH1)which is an epithelial cell adhesion molecule.

Western Blot:NIH/3T3 Cells, 1:200-1:2000

Type: Primary
Antigen: FZR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR