Anti-GUSB Rabbit Polyclonal Antibody

Supplier: Proteintech
16332-1-AP-150UL
10088-000EA 402 USD
10088-000
Anti-GUSB Rabbit Polyclonal Antibody
Antibodies
The GUSB gene encodes beta-glucuronidase, a lysosomal hydrolase involved in the stepwise degradation of glucuronic acid-containing glycosaminoglycans.It is a tetrameric glycoprotein composed of identical subunits.GUSB plays an important role in the degradation of dermatan and keratan sulfates.It has two isoforms.

Western Blot: Mouse Liver Tissue, 1:200-1:2000; IHC: Human Colon cancer Tissue, 1:20-1:200

Type: Primary
Antigen: GUSB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR