Anti-dGK Rabbit Polyclonal Antibody

Supplier: Proteintech
16314-1-AP-150UL
10085-802EA 402 USD
10085-802
Anti-dGK Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Kidney Tissue, 1:200-1:2000; IHC: Human hepatocirrhosis Tissue, 1:20-1:200

Type: Primary
Antigen: dGK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR