Anti-FN3KRP Rabbit Polyclonal Antibody

Supplier: Proteintech
16312-1-AP-150UL
10087-088EA 402 USD
10087-088
Anti-FN3KRP Rabbit Polyclonal Antibody
Antibodies
IHC: Human Brain Tissue, 1:20-1:200; Western Blot: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: FN3KRP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR