Anti-ATP5L Rabbit Polyclonal Antibody

Supplier: Proteintech
16307-1-AP-150UL
10083-350EA 402 USD
10083-350
Anti-ATP5L Rabbit Polyclonal Antibody
Antibodies
Mitochondrial membrane ATP synthase (F1-Fo ATP synthase or Complex V) produces ATP from ADP in the presence of a proton gradient across the membrane which is generated by electron transport complexes of the respiratory chain. It is composed of the soluble catalytic core, F1, and the membrane-spanning component and Fo, which comprises the proton channel. The Fo seems to have nine subunits (a, b, c, d, e, f, g, F6 and 8). ATP5L gene encodes ATP synthase subunit g of the Fo complex.

Western Blot: Mouse Liver Tissue, 1:500-1:5000; IHC: Human Kidney, 1:20-1:200

Type: Primary
Antigen: ATP5L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR