Anti-CHAC2 Rabbit Polyclonal Antibody

Supplier: Proteintech
16304-1-AP-150UL
10084-418EA 402 USD
10084-418
Anti-CHAC2 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:500-1:5000; IF: MCF-7 Cells, 1:10-1:100

Type: Primary
Antigen: CHAC2
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR