Anti-ARC Rabbit Polyclonal Antibody

Supplier: Proteintech
16290-1-AP-150UL
10082-302EA 402 USD
10082-302
Anti-ARC Rabbit Polyclonal Antibody
Antibodies
ARC, also named as KIAA0278 and Arg3.1, mediates endocytosis of neuronal AMPA-type glutamate receptors (AMPARs). It is required for consolidation of synaptic plasticity as well as formation of long-term memory. ARC plays a role in the regulation of cell morphology and cytoskeletal organization. It is required in the stress fiber dynamics and cell migration.

Western Blot:SH-SY5Y Cells, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: ARC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR