Anti-CHMP6 Rabbit Polyclonal Antibody

Supplier: Proteintech
16278-1-AP-150UL
10084-722EA 402 USD
10084-722
Anti-CHMP6 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Testis Tissue, 1:200-1:2000

Type: Primary
Antigen: CHMP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR