Anti-DHRS1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16275-1-AP-150UL
10085-818EA 402 USD
10085-818
Anti-DHRS1 Rabbit Polyclonal Antibody
Antibodies
DHRS1(Dehydrogenase/reductase SDR family member 1) is also named as LJ25430, MGC20204, DKFZp586I0523, SDR19C1, FLJ14250 and belongs to the short-chain dehydrogenases/reductases (SDR) family. It is involved in the metabolism of a large variety of compounds, including steroid hormones, prostaglandins, retinoids, lipids and xenobiotics.Northern blot analysis detected DHRS1express in heart highest and in liver lowest.

Western Blot:COLO 320 Cells, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: DHRS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR