Anti-SYAP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16272-1-AP-150UL
10095-484EA 402 USD
10095-484
Anti-SYAP1 Rabbit Polyclonal Antibody
Antibodies
SYAP1(Synapse-associated protein 1) is a human homologue of the Drosophila SAP47 (synapse associated protein), which is recognized by a monoclonal antibody that selectively stain synaptic terminals.This protein is a 352 amino acid protein that is ubiquitously expressed in adult tissues. SYAP1 contains one BSD domain which is is an approximately 60 amino acid long protein domain named after the BTF2-like transcription factors, Synapse-associated proteins and DOS2-like proteins in which it is found.

Western Blot: HEK-293 Cells, 1:200-1:2000; IHC: Human cervical cancer Tissue, 1:20-1:200

Type: Primary
Antigen: SYAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR