Anti-MED28 Rabbit Polyclonal Antibody

Supplier: Proteintech
16262-1-AP-150UL
10090-096EA 402 USD
10090-096
Anti-MED28 Rabbit Polyclonal Antibody
Antibodies
IHC: Human Testis, 1:20-1:200

Type: Primary
Antigen: MED28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR