Anti-GADD45GIP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16260-1-AP-150UL
10087-310EA 402 USD
10087-310
Anti-GADD45GIP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Heart Tissue, 1:200-1:2000; IP: Mouse Heart Tissue, 1:200-1:1000

Type: Primary
Antigen: GADD45GIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR