Anti-CAP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16231-1-AP-150UL
10084-048EA 402 USD
10084-048
Anti-CAP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse skin Tissue, 1:200-1:2000; IHC: Human pancreas cancer Tissue, 1:20-1:200; FC: HeLa Cells, N/A

Type: Primary
Antigen: CAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR