Anti-MKS1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16206-1-AP-150UL
10090-258EA 402 USD
10090-258
Anti-MKS1 Rabbit Polyclonal Antibody
Antibodies
MKS1 (Meckel syndrome type 1 protein) is a 559-amino acid protein that contains a conserved B9 domain. It is a component of a large protein complex which localizes to the ciliary transition zone and regulates mammalian ciliogenesis and ciliary membrane composition . MKS1 is required for ciliary structure and function, and is involved in centrosome migration to the apical cell surface during early ciliogenesis . Broad tissue expression of the MKS1 gene has been reported . Defects in MKS1 are the cause of Meckel syndrome type 1 (MKS1), an autosomal recessive lethal malformation syndrome characterized by renal cystic dysplasia, central nervous system malformations, and hepatic developmental defects . In addition, defects in MKS1 are also the cause of Bardet-Biedl syndrome type 13 (BBS13) .

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human Liver cancer Tissue, 1:20-1:200; IF: HTERT-RPE1 Cells and Mouse Embryonic fibroblasts, 1:20-1:200; IP: HEK-293 Cells, 1:500-1:5000

Type: Primary
Antigen: MKS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR