Anti-ING3 Rabbit Polyclonal Antibody

Supplier: Proteintech
16187-1-AP-150UL
10088-890EA 402 USD
10088-890
Anti-ING3 Rabbit Polyclonal Antibody
Antibodies
Members of inhibitor of growth (ING) family function in inhibiting cell growth and inducing apoptosis. They are sequence homologous proteins. ING3 can activate p53 trans-activated promoters, including promoters of p21/waf1 and bax. This antibody is specifically against p47ING3.

Western Blot: Human Kidney Tissue, 1:500-1:5000

Type: Primary
Antigen: ING3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR