Anti-ICAM1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16174-1-AP-150UL
10088-182EA 453 USD
10088-182
Anti-ICAM1 Rabbit Polyclonal Antibody
Antibodies
ICAM-1 (Intercellular Adhesion Molecule-1, CD54), a member of Ig super family, is a glycosylated, transmembrane protein . This protein is involved in cell adhesion by interacting with a number of integrin family members such as LFA-1 and Mac-1. Its expression is regulated by chemokines. This protein migrates approximately 115 kDa on SDS gels. This antibody is specifically againt mouse ICAM1.

IF:4T1 Cells, 1:20-1:200; Western Blot:rat Heart Tissue, 1:200-1:2000; IHC: Mouse lung Tissue, 1:20-1:200

Type: Primary
Antigen: ICAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR