Anti-HDAC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16160-1-AP-150UL
10088-122EA 402 USD
10088-122
Anti-HDAC1 Rabbit Polyclonal Antibody
Antibodies
Histone deacetylases(HDAC) are a class of enzymes that remove the acetyl groups from the lysine residues leading to the formation of a condensed and transcriptionally silenced chromatin. The protein encoded by this gene belongs to the histone deacetylase/acuc/apha family and is a component of the histone deacetylase complex, which is responsible for gene expression silencing. It also plays an important role in the control of cell proliferation and differentiation by interacting with RB, p53 and other transcription factors. At least 4 classes of HDAC were identified. As a class I HDAC, HDAC 1 was primarily found in the nucleus. This antibody is a rabbit polyclonal antibody raised against a peptide mapping near the N-terminus of human HDAC1 and is speicifc to HDAC1. It does not react with other HDACs.

Western Blot: Mouse Testis Tissue, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: HDAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR