Anti-KRAS-2A Rabbit Polyclonal Antibody

Supplier: Proteintech
16156-1-AP-150UL
10081-600EA 537.7 USD
10081-600
Anti-KRAS-2A Rabbit Polyclonal Antibody
Antibodies
RAS (derived from 'RAt Sarcoma' virus) gene family includes KRAS, HRAS, and NRAS. These genes encode very similar 21KD proteins and are homologs of rodent sarcoma virus genes that have transforming abilities. KRAS has two isoforms, a and b. This antibody is specific for KRASA.

Western Blot:SH-SY5Y Cells, 1:200-1:2000; IHC: Human lung cancer Tissue, 1:20-1:200; FC: HeLa Cells, N/A

Type: Primary
Antigen: KRAS-2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR