Anti-KRAS-2B Rabbit Polyclonal Antibody

Supplier: Proteintech
16155-1-AP-150UL
10081-598EA 537.7 USD
10081-598
Anti-KRAS-2B Rabbit Polyclonal Antibody
Antibodies
RAS gene family encodes very similar 21KD proteins and are homologs of rodent sarcoma virus genes that have transforming abilities. KRAS has two isoforms, a and b. This antibody is specific for KRASB.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human pancreas cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; FC: HepG2 Cells, N/A

Type: Primary
Antigen: KRAS-2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR