Anti-POLR1E Rabbit Polyclonal Antibody

Supplier: Proteintech
16145-1-AP-150UL
10092-570EA 402 USD
10092-570
Anti-POLR1E Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:1000-1:10000; IP:Jurkat Cells, 1:200-1:2000

Type: Primary
Antigen: POLR1E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR