Anti-Citrate synthase Rabbit Polyclonal Antibody

Supplier: Proteintech
16131-1-AP-150UL
10085-230EA 402 USD
10085-230
Anti-Citrate synthase Rabbit Polyclonal Antibody
Antibodies
Citrate synthase (CS), the first and rate-limiting enzyme of the tricarboxylic acid cycle, plays a key role in regulating energy generation of mitochondrial respiration.It belongs to the citrate synthase family. The deduced 466-amino acid protein contains an N-terminal mitochondrial targeting sequence and a motif highly conserved in citrate synthases. It can exsit as a dimer. Northern blot analysis detected no CS expression in thymus and small intestine. This antibody is specific to CS.

Western Blot: HEK-293 Cells, 1:1000-1:10000; IHC: Human Heart Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: Citrate synthase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR