Anti-NXN Rabbit Polyclonal Antibody

Supplier: Proteintech
16128-1-AP-150UL
10091-504EA 402 USD
10091-504
Anti-NXN Rabbit Polyclonal Antibody
Antibodies
Nucleoredoxin (NRX/NXN), a relatively uncharacterized member of the TRX family protein, has recently been reported to regulate the Wnt/β-catenin pathway, which itself regulates cell fate and early development, in a redox-dependent manner. Northern blot analysis detected several nucleoredoxin splice variants in adult mouse tissues, with highest expression in testis and skin and lowest expression in liver. It has 3 isoforms produced by alternative splicing. This antibody is specific to NXN.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Testis, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: NXN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR