Anti-RAC1/2/3 Rabbit Polyclonal Antibody

Supplier: Proteintech
16117-1-AP-150UL
10093-264EA 402 USD
10093-264
Anti-RAC1/2/3 Rabbit Polyclonal Antibody
Antibodies
Rac3 is a GTPase that belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. There are three Rac genes in mammals, which share over 90% protein identity but differ for their pattern of expression. Rac1 is ubiquitous, Rac2 is specific for hematopoietic cells, while Rac3 is prevalent in the developing nervous system. This antibody can recognize Rac1, Rac2, and Rac3.

Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human Brain, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: RAC1/2/3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR