Anti-GS28 Rabbit Polyclonal Antibody

Supplier: Proteintech
16106-1-AP-150UL
10087-726EA 402 USD
10087-726
Anti-GS28 Rabbit Polyclonal Antibody
Antibodies
GS28, also known as GOSR1, is a 28-kDa Golgi SNARE protein that participates in ER-Golgi and intra-Golgi transport. GS28 is considered to be a core component of the Golgi SNARE complex.

Western Blot: Human Liver Tissue, 1:500-1:5000; IHC: Human Stomach cancer Tissue, 1:20-1:200; FC: HeLa Cells, N/A

Type: Primary
Antigen: GS28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR