Anti-S100 A4 Rabbit Polyclonal Antibody

Supplier: Proteintech
16105-1-AP-150UL
10094-086EA 402 USD
10094-086
Anti-S100 A4 Rabbit Polyclonal Antibody
Antibodies
S100A4 is a member of the S100 family of calcium-binding proteins. The S100 family members have been involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100A4 is known to localize to and function in the nucleus, cytoplasm of cells, and the extracellular space. S100A4 has also been shown to be associated with tumor growth, motility, invasion, metastasis, angiogenesis, apoptosis, and chemoresistance. Overexpression of S100A4 is correlated with a worse prognosis inpatients with various types of cancer.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: S100 A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR