Anti-GNB2 Rabbit Polyclonal Antibody

Supplier: Proteintech
16090-1-AP-150UL
10087-692EA 402 USD
10087-692
Anti-GNB2 Rabbit Polyclonal Antibody
Antibodies
GNB2(Guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-2) is also named as G protein subunit beta-2, transducin beta chain 2 and belongs to the WD repeat G protein beta family. G proteins are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human ovary tumor Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: GNB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR