Anti-GCDFP15 Rabbit Polyclonal Antibody

Supplier: Proteintech
16068-1-AP-150UL
10081-794EA 402 USD
10081-794
Anti-GCDFP15 Rabbit Polyclonal Antibody
Antibodies
GCDFP-15 (gross cystic disease fluid protein 15), also known as PIP (prolactin-induced protein) is a secretory glycoprotein expressed in benign and malignant breast tumor tissues and in some normal exocrine organs such as sweat, salivary, and lacrimal glands . GCDFP-15 expression is increased by prolactin and androgen and inhibited by estrogen . The expression is also regulated by interleukins. GCDFP-15 is a marker of apocrine differentiation and is frequently used for assessment of metastases or regional recurrences of breast origin.

IHC: Human skin Tissue, 1:20-1:200

Type: Primary
Antigen: GCDFP-15/PIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR