Anti-PMPCB Rabbit Polyclonal Antibody

Supplier: Proteintech
16064-1-AP-150UL
10092-298EA 402 USD
10092-298
Anti-PMPCB Rabbit Polyclonal Antibody
Antibodies
PMPCB(Mitochondrial-processing peptidase subunit beta) is also named as MPPB, P-52 and belongs to the peptidase M16 family. It cleaves presequences (transit peptides) from mitochondrial protein precursors. It is a mitochondrial matrix marker and Tom70, cytochrome c heme lyase (CCHL) are markers for outer membrane, intermembrane space.

Western Blot: MCF7 Cells, 1:500-1:5000; IHC: Human lung cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP: MCF-7 Cells, 1:200-1:2000

Type: Primary
Antigen: PMPCB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR