Anti-PDCL Rabbit Polyclonal Antibody

Supplier: Proteintech
16057-1-AP-150UL
10091-986EA 402 USD
10091-986
Anti-PDCL Rabbit Polyclonal Antibody
Antibodies
  • Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human Placenta, 1:20-1:200

Type: Primary
Antigen: PDCL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR