Anti-ATP6V1C1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16054-1-AP-150UL
10083-366EA 402 USD
10083-366
Anti-ATP6V1C1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:NIH/3T3 Cells, 1:500-1:5000; IHC: Human Stomach cancer, 1:20-1:200

Type: Primary
Antigen: ATP6V1C1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR