Anti-SRP19 Rabbit Polyclonal Antibody

Supplier: Proteintech
16033-1-AP-150UL
10095-170EA 402 USD
10095-170
Anti-SRP19 Rabbit Polyclonal Antibody
Antibodies
The signal recognition particle (SRP) is one of the few functional small RNP particles. The SRP couples the synthesis of membrane and secretory proteins across or into the endoplasmic reticulum (ER) membrane in eukaryotes, as well as across the bacterial plasma membrane, and chloroplast thylakoid membranes. The mammalian SRP is composed of a 7S (or 7SL) RNA and six different proteins, SRP9, SRP14, SRP19, SRP54, SRP68 and SRP72. All of the components of SRP, including SRP RNA, participate directly in the overall protein targeting process. SRP19 binds directly to 7S RNA and mediates binding of the 54 kDa subunit of the SRP. SRP19 was shown to significantly enhance SRP54 attachment to helix 8 of 7SL RNA. Binding of SRP19 leads to restructuring of both helix 6 and 8, causing local changes at the SRP54-binding site. This antibody is a rabbit polyclonal antibody raised against full length SRP19 of human origin.

Western Blot: Human Liver Tissue, 1:500-1:5000

Type: Primary
Antigen: SRP19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR