Anti-MMS19 Rabbit Polyclonal Antibody

Supplier: Proteintech
16015-1-AP-150UL
10090-316EA 402 USD
10090-316
Anti-MMS19 Rabbit Polyclonal Antibody
Antibodies
MMS19 plays a critical role in the biogenesis of multiple Fe-S proteins, which functions in genomic stability, RNA polymerase II function, and telomere length regulation. MMS19 has a alanine- and leucine-rich N terminus. It possible has a role in nucleotide excision repair(NER) and RNA polymerase II(POL II) transcription by interacting with ERCC2/XPB helicases, both subunits of NER-transcription factor TFIIH. Also MMS19 involves in the regualtion of ER. It can function in chromosome segregation by acting as a part of the mitotic spindle-associated MMXD complex.

Western Blot: Human Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: MMS19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR