Anti-TRAPPC9 Rabbit Polyclonal Antibody

Supplier: Proteintech
16014-1-AP-150UL
10096-290EA 402 USD
10096-290
Anti-TRAPPC9 Rabbit Polyclonal Antibody
Antibodies
TRAPPC9, also named as KIAA1882 and NIBP, belongs to the NIBP family. It functions as an activator of NF-kappa-B through increased phosphorylation of the IKK complex. TRAPPC9 may function in neuronal cells differentiation and play a role in vesicular transport from endoplasmic reticulum to Golgi. TRAPPC9 was found in neurons of the cerebral cortex, hippocampus, and deep gray matter. Western blotting indicates molecular weight of TRAPPC9 is 130-140 kDa, and 250 kDa may be detected with extensive posttranslational modification of TRAPPC9.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: TRAPPC9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR